missing translation for 'onlineSavingsMsg'
Learn More

5-Benzyloxyindole 98.0+%, TCI America™

Supplier:  TCI America B18335G

Catalog No. B18335G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

5-Benzyloxyindole
C15H13NO
5 g
JCQLPDZCNSVBMS-UHFFFAOYSA-N
5-phenylmethoxy-1H-indole
14624
≥98.0% (HPLC,N)
1215-59-4
MFCD00005676
5-benzyloxyindole, 5-benzyloxy-1h-indole, 1h-indole, 5-phenylmethoxy, benzyloxy-5 indole, indole, 5-benzyloxy, 5-benzyloxy indole, unii-yci4z02e1c
C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3
223.275
223.28
Crystalline Powder

Chemical Identifiers

1215-59-4
223.275
JCQLPDZCNSVBMS-UHFFFAOYSA-N
14624
C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3
C15H13NO
MFCD00005676
5-benzyloxyindole, 5-benzyloxy-1h-indole, 1h-indole, 5-phenylmethoxy, benzyloxy-5 indole, indole, 5-benzyloxy, 5-benzyloxy indole, unii-yci4z02e1c
5-phenylmethoxy-1H-indole
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : NL4850000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions