missing translation for 'onlineSavingsMsg'
Learn More

5-Benzyloxyindole, 95%, Thermo Scientific™

Chemical Identifiers

CAS 1215-59-4
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.27
MDL Number MFCD00005676
InChI Key JCQLPDZCNSVBMS-UHFFFAOYSA-N
Synonym 5-benzyloxyindole, 5-benzyloxy-1h-indole, 1h-indole, 5-phenylmethoxy, benzyloxy-5 indole, indole, 5-benzyloxy, 5-benzyloxy indole, unii-yci4z02e1c
PubChem CID 14624
IUPAC Name 5-phenylmethoxy-1H-indole
SMILES C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC106090050
SDS View Documents
Thermo Scientific Chemicals
106090050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC106090250
SDS View Documents
Thermo Scientific Chemicals
106090250
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1215-59-4
223.27
JCQLPDZCNSVBMS-UHFFFAOYSA-N
14624
C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3
C15H13NO
MFCD00005676
5-benzyloxyindole, 5-benzyloxy-1h-indole, 1h-indole, 5-phenylmethoxy, benzyloxy-5 indole, indole, 5-benzyloxy, 5-benzyloxy indole, unii-yci4z02e1c
5-phenylmethoxy-1H-indole

Specifications

1215-59-4
100.0
6% max. Toluene (GC)
C15H13NO
5g
Solubility in water: practically insoluble in water. Other solubilities: soluble in chloroform and (warm) ethanol
C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3
223.27
223.27
5-Benzyloxyindole, 95%
94.0
Authentic
Glass bottle
MFCD00005676
5-benzyloxyindole, 5-benzyloxy-1h-indole, 1h-indole, 5-phenylmethoxy, benzyloxy-5 indole, indole, 5-benzyloxy, 5-benzyloxy indole, unii-yci4z02e1c
JCQLPDZCNSVBMS-UHFFFAOYSA-N
5-phenylmethoxy-1H-indole
14624
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 214-930-1

RTECSNumber : NL4850000

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only