missing translation for 'onlineSavingsMsg'
Learn More

5-Benzyloxyindole, 94%, may contain up to ca 7% toluene

CAS: 1215-59-4 | C15H13NO | 223.275 g/mol

Supplier:  Thermo Scientific Chemicals B2514306

 View more versions of this product

Catalog No. AAB2514306


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Benzyloxyindole
96°C to 101°C
C15H13NO
5 g
5-benzyloxyindole, 5-benzyloxy-1h-indole, 1h-indole, 5-phenylmethoxy, benzyloxy-5 indole, indole, 5-benzyloxy, 5-benzyloxy indole, unii-yci4z02e1c
C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3
223.275
223.28
1215-59-4
may contain up to ≈7% toluene
MFCD00005676
173532
JCQLPDZCNSVBMS-UHFFFAOYSA-N
5-phenylmethoxy-1H-indole
14624
94%

Chemical Identifiers

1215-59-4
223.275
JCQLPDZCNSVBMS-UHFFFAOYSA-N
14624
C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3
C15H13NO
MFCD00005676
5-benzyloxyindole, 5-benzyloxy-1h-indole, 1h-indole, 5-phenylmethoxy, benzyloxy-5 indole, indole, 5-benzyloxy, 5-benzyloxy indole, unii-yci4z02e1c
5-phenylmethoxy-1H-indole
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P331-P332+P313-P362-P501c

H302+H312+H332-H304-H315-H319-H335-H336-H361d

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 214-930-1

RTECSNumber : NL4850000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only