missing translation for 'onlineSavingsMsg'
Learn More

5-Benzyloxyindole-3-carboxaldehyde, 98%

Catalog No. AAL0111703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0111703 1 g
AAL0111706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0111703 Supplier Thermo Scientific Chemicals Supplier No. L0111703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6953-22-6 | C16H13NO2 | 251.285 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6953-22-6
Molecular Formula C16H13NO2
Molecular Weight (g/mol) 251.285
MDL Number MFCD00014562
InChI Key DJGNUBADRQIDNQ-UHFFFAOYSA-N
Synonym 5-benzyloxyindole-3-carboxaldehyde, 5-benzyloxyindole-3-carbaldehyde, 5-benzyloxy-1h-indole-3-carbaldehyde, 5-phenylmethoxy-1h-indole-3-carbaldehyde, 5-benzyloxy-3-formylindole, 5-benzyloxy-indole-3-carboxaldehyde, 5-benzyloxy-3-formyl-1h-indole, 5-benzyloxy-3-indolecarboxaldehyde, 5-benzyloxy indole-3-carboxyaldehyde
PubChem CID 81398
IUPAC Name 5-phenylmethoxy-1H-indole-3-carbaldehyde
SMILES C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC=C3C=O

Specifications

Melting Point 239°C to 242°C
Quantity 1 g
Beilstein 208873
Sensitivity Air Sensitive
Formula Weight 251.28
Percent Purity 98%
Chemical Name or Material 5-Benzyloxyindole-3-carboxaldehyde
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 230-134-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only