missing translation for 'onlineSavingsMsg'
Learn More

5-Benzyl N-Carbobenzoxy-L-glutamate 98.0+%, TCI America™

Catalog No. B39025G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B39025G Supplier TCI America Supplier No. B39025G
Only null left

Specifications

CAS 5680-86-4
Melting Point 76°C
Color White
Molecular Formula C20H21NO6
Quantity 5 g
MDL Number MFCD00065697
Synonym z-glu obzl-oh, cbz-glu obzl-oh, 2s-5-benzyloxy-2-benzyloxy carbonyl amino-5-oxopentanoic acid, 5-benzyl n-carbobenzoxy-l-glutamate, n-cbz-l-glutamic acid 5-benzyl ester, s-2-benzyloxycarbonylamino-pentanedioic acid 5-benzyl ester, s-5-benzyloxy-2-benzyloxy carbonyl amino-5-oxopentanoic acid, z-glu obzl oh, pubchem14964, z-glu-obzl-oh
InChI Key TWIVXHQQTRSWGO-KRWDZBQOSA-N
SMILES C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)NC(=O)OCC2=CC=CC=C2
IUPAC Name (2S)-5-oxo-5-phenylmethoxy-2-(phenylmethoxycarbonylamino)pentanoic acid
Molecular Weight (g/mol) 371.389
PubChem CID 3049860
Formula Weight 371.39
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Benzyl N-Carbobenzoxy-L-glutamate
Show More Show Less