missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Benzyl D-Glutamate 98.0+%, TCI America™
Supplier: TCI America B39995G
Specifications
| 5-Benzyl D-Glutamate | |
| 158°C | |
| C12H15NO4 | |
| 5 g | |
| BGGHCRNCRWQABU-SNVBAGLBSA-N | |
| (2R)-2-amino-5-oxo-5-phenylmethoxypentanoic acid | |
| 6994624 | |
| ≥98.0% (HPLC,T) |
| 2578-33-8 | |
| White | |
| MFCD00063111 | |
| h-d-glu obzl-oh, d-glutamic acid 5-benzyl ester, d-glutamic acid gamma-benzyl ester, 2r-2-amino-5-benzyloxy-5-oxopentanoic acid, 2r-2-amino-5-oxo-5-phenylmethoxypentanoic acid, 2r-2-amino-5-oxo-5-phenylmethoxypentanoic acid, d-glutamicacid5-benzylester, h-d-glu oblz-oh, h-d-glu oblz-oh, a-benzyl ester, a-benzyl ester | |
| C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)N | |
| 237.255 | |
| 237.26 | |
| Crystalline Powder |
Chemical Identifiers
| 2578-33-8 | |
| 237.255 | |
| BGGHCRNCRWQABU-SNVBAGLBSA-N | |
| 6994624 | |
| C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)N |
| C12H15NO4 | |
| MFCD00063111 | |
| h-d-glu obzl-oh, d-glutamic acid 5-benzyl ester, d-glutamic acid gamma-benzyl ester, 2r-2-amino-5-benzyloxy-5-oxopentanoic acid, 2r-2-amino-5-oxo-5-phenylmethoxypentanoic acid, 2r-2-amino-5-oxo-5-phenylmethoxypentanoic acid, d-glutamicacid5-benzylester, h-d-glu oblz-oh, h-d-glu oblz-oh, a-benzyl ester, a-benzyl ester | |
| (2R)-2-amino-5-oxo-5-phenylmethoxypentanoic acid |
Safety and Handling
TSCA : No