missing translation for 'onlineSavingsMsg'
Learn More

5-[(Benzoylamino)methyl]thiophene-2-sulfonyl chloride, 97%, Thermo Scientific™

Catalog No. KM04426CB
Change view
Click to view available options
Quantity:
250mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
KM04426CB 250mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KM04426CB Supplier Thermo Scientific Chemicals Supplier No. KM04426CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 138872-44-3
Molecular Formula C12H10ClNO3S2
Molecular Weight (g/mol) 315.786
MDL Number MFCD00052130
InChI Key VGSWVDWOXYTAPG-UHFFFAOYSA-N
Synonym 5-benzoylamino methyl thiophene-2-sulfonyl chloride, 5-benzamidomethyl thiophene-2-sulfonyl chloride, 2-benzoylaminomethyl thiophene-5-sulfonylchloride, 2-benzoylaminomethyl thiophene-5-sulfonyl chloride, 2-thiophenesulfonylchloride, 5-benzoylamino methyl, 5-phenylformamido methyl thiophene-2-sulfonyl chloride, 5-benzoylamino methyl thiophene-2-sulphonyl chloride, pubchem5470, acmc-20c3wu, 5-benzamidomethyl thiophene-2-sulfonylchloride
PubChem CID 2778878
IUPAC Name 5-(benzamidomethyl)thiophene-2-sulfonyl chloride
SMILES C1=CC=C(C=C1)C(=O)NCC2=CC=C(S2)S(=O)(=O)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 315.8
Percent Purity 97%
Chemical Name or Material 5-[(Benzoylamino)methyl]thiophene-2-sulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.