missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Benzimidazolecarboxylic Acid 97.0+%, TCI America™
$128.62 - $512.62
Chemical Identifiers
| CAS | 15788-16-6 |
|---|---|
| Molecular Formula | C8H6N2O2 |
| Molecular Weight (g/mol) | 162.15 |
| MDL Number | MFCD00011555 |
| InChI Key | COYPLDIXZODDDL-UHFFFAOYSA-N |
| Synonym | 1h-benzimidazole-5-carboxylic acid, 5-benzimidazolecarboxylic acid, benzimidazole-5-carboxylic acid, 1h-benzo d imidazole-5-carboxylic acid, 1h-benzimidazole-6-carboxylic acid, 5-carboxybenzimidazole, 1h-1,3-benzodiazole-5-carboxylic acid, 3h-benzo d imidazole-5-carboxylic acid, 1h-benzimidazole-5-carboxylicacid, 1h-1,3-benzodiazole-6-carboxylic acid |
| PubChem CID | 459456 |
| IUPAC Name | 1H-1,3-benzodiazole-6-carboxylic acid |
| SMILES | OC(=O)C1=CC=C2N=CNC2=C1 |
Chemical Identifiers
| 15788-16-6 | |
| 162.15 | |
| COYPLDIXZODDDL-UHFFFAOYSA-N | |
| 459456 | |
| OC(=O)C1=CC=C2N=CNC2=C1 |
| C8H6N2O2 | |
| MFCD00011555 | |
| 1h-benzimidazole-5-carboxylic acid, 5-benzimidazolecarboxylic acid, benzimidazole-5-carboxylic acid, 1h-benzo d imidazole-5-carboxylic acid, 1h-benzimidazole-6-carboxylic acid, 5-carboxybenzimidazole, 1h-1,3-benzodiazole-5-carboxylic acid, 3h-benzo d imidazole-5-carboxylic acid, 1h-benzimidazole-5-carboxylicacid, 1h-1,3-benzodiazole-6-carboxylic acid | |
| 1H-1,3-benzodiazole-6-carboxylic acid |
Specifications
| 15788-16-6 | |
| MFCD00011555 | |
| 1h-benzimidazole-5-carboxylic acid, 5-benzimidazolecarboxylic acid, benzimidazole-5-carboxylic acid, 1h-benzo d imidazole-5-carboxylic acid, 1h-benzimidazole-6-carboxylic acid, 5-carboxybenzimidazole, 1h-1,3-benzodiazole-5-carboxylic acid, 3h-benzo d imidazole-5-carboxylic acid, 1h-benzimidazole-5-carboxylicacid, 1h-1,3-benzodiazole-6-carboxylic acid | |
| OC(=O)C1=CC=C2N=CNC2=C1 | |
| 162.15 | |
| 162.15 | |
| Crystalline Powder |
| C8H6N2O2 | |
| 5 g | |
| COYPLDIXZODDDL-UHFFFAOYSA-N | |
| 1H-1,3-benzodiazole-6-carboxylic acid | |
| 459456 | |
| ≥97.0% (T) | |
| 5-Benzimidazolecarboxylic Acid |
Safety and Handling
TSCA : No