missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Benzimidazolecarboxylic Acid 97.0+%, TCI America™
Specifications
Specifications
| Quantity | 5 g |
| Formula Weight | 162.15 |
| Percent Purity | ≥97.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-Benzimidazolecarboxylic Acid |
Safety and Handling
| TSCA | No |