missing translation for 'onlineSavingsMsg'
Learn More

5-Azoniaspiro[4.4]nonane Iodide 98.0+%, TCI America™

Catalog No. p-7118217
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30931G 1 g
A30935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A30931G Supplier TCI America Supplier No. A30931G
Only null left

Chemical Identifiers

CAS 45650-35-9
Molecular Formula C8H16IN
Molecular Weight (g/mol) 253.127
InChI Key DWOWCUCDJIERQX-UHFFFAOYSA-M
PubChem CID 86209376
IUPAC Name 5-azoniaspiro[4.4]nonane;iodide
SMILES C1CC[N+]2(C1)CCCC2.[I-]

Specifications

Color White
Quantity 1 g
Formula Weight 253.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Azoniaspiro[4.4]nonane Iodide