missing translation for 'onlineSavingsMsg'
Learn More

5-Azoniaspiro[4.4]nonane Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

Catalog No. A31011G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31011G 1 g
A31015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A31011G Supplier TCI America Supplier No. A31011G

Chemical Identifiers

CAS 706759-44-6
Molecular Formula C10H16F6N2O4S2

Specifications

Melting Point 114°C
Color White
Quantity 1 g
Formula Weight 406.36
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Azoniaspiro[4.4]nonane Bis(trifluoromethanesulfonyl)imide