missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Azacytosine, 98% (dry wt.), may cont. up to ca 7% water
CAS: 931-86-2 | C3H4N4O | 112.09 g/mol
$78.03 - $259.49
Chemical Identifiers
| CAS | 931-86-2 |
|---|---|
| Molecular Formula | C3H4N4O |
| Molecular Weight (g/mol) | 112.09 |
| MDL Number | MFCD00006033,MFCD00149402,MFCD00051007 |
| InChI Key | MFEFTTYGMZOIKO-UHFFFAOYSA-N |
| Synonym | 5-azacytosine, 1,3,5-triazin-2 1h-one, 4-amino, 4-amino-1,3,5-triazin-2 1h-one, 4-amino-1,3,5-triazin-2-ol, 4-amino-1,3,5-triazin-2-one, s-triazin-2 1h-one, 4-amino, s-triazin-2-ol, 4-amino, 2-amino-4-hydroxy-s-triazine, 6-amino-1,3,5-triazin-2 1h-one, 4-amino-s-triazin-2 1h-one |
| PubChem CID | 19956 |
| ChEBI | CHEBI:72474 |
| SMILES | NC1=NC=NC(=O)N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1341006
|
Thermo Scientific Chemicals
A1341006 |
5 g |
Each for $78.03
|
|
|||||
|
AAA1341014
|
Thermo Scientific Chemicals
A1341014 |
25 g |
Each for $259.49
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 931-86-2 | |
| 112.09 | |
| MFEFTTYGMZOIKO-UHFFFAOYSA-N | |
| 19956 | |
| NC1=NC=NC(=O)N1 |
| C3H4N4O | |
| MFCD00006033,MFCD00149402,MFCD00051007 | |
| 5-azacytosine, 1,3,5-triazin-2 1h-one, 4-amino, 4-amino-1,3,5-triazin-2 1h-one, 4-amino-1,3,5-triazin-2-ol, 4-amino-1,3,5-triazin-2-one, s-triazin-2 1h-one, 4-amino, s-triazin-2-ol, 4-amino, 2-amino-4-hydroxy-s-triazine, 6-amino-1,3,5-triazin-2 1h-one, 4-amino-s-triazin-2 1h-one | |
| CHEBI:72474 |
Specifications
| 931-86-2 | |
| C3H4N4O | |
| 5 g | |
| 5-azacytosine, 1,3,5-triazin-2 1h-one, 4-amino, 4-amino-1,3,5-triazin-2 1h-one, 4-amino-1,3,5-triazin-2-ol, 4-amino-1,3,5-triazin-2-one, s-triazin-2 1h-one, 4-amino, s-triazin-2-ol, 4-amino, 2-amino-4-hydroxy-s-triazine, 6-amino-1,3,5-triazin-2 1h-one, 4-amino-s-triazin-2 1h-one | |
| NC1=NC=NC(=O)N1 | |
| 112.09 | |
| CHEBI:72474 | |
| 98% | |
| (dry wt.), may cont. up to ≈7% water |
| >300°C | |
| MFCD00006033,MFCD00149402,MFCD00051007 | |
| 116378 | |
| MFEFTTYGMZOIKO-UHFFFAOYSA-N | |
| 6-amino-1H-1,3,5-triazin-2-one | |
| 19956 | |
| 112.09 | |
| 5-Azacytosine |
Safety and Handling
EINECSNumber : 213-242-9
RTECSNumber : XZ2854300
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only