missing translation for 'onlineSavingsMsg'
Learn More

5-Azacytosine, 98% (dry wt.), may cont. up to ca 7% water

Catalog No. AAA1341006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1341006 5 g
AAA1341014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1341006 Supplier Thermo Scientific Chemicals Supplier No. A1341006

CAS: 931-86-2 | C3H4N4O | 112.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 931-86-2
Molecular Formula C3H4N4O
Molecular Weight (g/mol) 112.09
MDL Number MFCD00006033,MFCD00149402,MFCD00051007
InChI Key MFEFTTYGMZOIKO-UHFFFAOYSA-N
Synonym 5-azacytosine, 1,3,5-triazin-2 1h-one, 4-amino, 4-amino-1,3,5-triazin-2 1h-one, 4-amino-1,3,5-triazin-2-ol, 4-amino-1,3,5-triazin-2-one, s-triazin-2 1h-one, 4-amino, s-triazin-2-ol, 4-amino, 2-amino-4-hydroxy-s-triazine, 6-amino-1,3,5-triazin-2 1h-one, 4-amino-s-triazin-2 1h-one
PubChem CID 19956
ChEBI CHEBI:72474
SMILES NC1=NC=NC(=O)N1

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 116378
IUPAC Name 6-amino-1H-1,3,5-triazin-2-one
Formula Weight 112.09
Percent Purity 98%
Assay (dry wt.), may cont. up to ≈7% water
Chemical Name or Material 5-Azacytosine
EINECSNumber 213-242-9
RTECSNumber XZ2854300
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only