missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Azabenzimidazole, 98%, Thermo Scientific Chemicals
$183.59 - $183.59
Chemical Identifiers
| CAS | 272-97-9 |
|---|---|
| Molecular Formula | C6H5N3 |
| Molecular Weight (g/mol) | 119.13 |
| MDL Number | MFCD00051808 |
| InChI Key | UBOOKRVGOBKDMM-UHFFFAOYSA-N |
| Synonym | 3h-imidazo 4,5-c pyridine, 1h-imidazo 4,5-c pyridine, 5-azabenzimidazole, 3,5-diazaindole, 5h-imidazo 4,5-c pyridine, imidazo 4,5-c pyridine, 5-aza-1h-benzoimidazole, 5-aza-benzimidazole |
| PubChem CID | 9227 |
| IUPAC Name | 3H-imidazo[4,5-c]pyridine |
| SMILES | N1C=NC2=CC=NC=C12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0693003
|
Thermo Scientific Chemicals
L0693003 |
1 g |
Each for $183.59
|
|
|||||
Description
5-Azabenzimidazole is used as Pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Azabenzimidazole is used as Pharmaceutical intermediates.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
Chemical Identifiers
| 272-97-9 | |
| 119.13 | |
| UBOOKRVGOBKDMM-UHFFFAOYSA-N | |
| 9227 | |
| N1C=NC2=CC=NC=C12 |
| C6H5N3 | |
| MFCD00051808 | |
| 3h-imidazo 4,5-c pyridine, 1h-imidazo 4,5-c pyridine, 5-azabenzimidazole, 3,5-diazaindole, 5h-imidazo 4,5-c pyridine, imidazo 4,5-c pyridine, 5-aza-1h-benzoimidazole, 5-aza-benzimidazole | |
| 3H-imidazo[4,5-c]pyridine |
Specifications
| 272-97-9 | |
| C6H5N3 | |
| 1 g | |
| 3h-imidazo 4,5-c pyridine, 1h-imidazo 4,5-c pyridine, 5-azabenzimidazole, 3,5-diazaindole, 5h-imidazo 4,5-c pyridine, imidazo 4,5-c pyridine, 5-aza-1h-benzoimidazole, 5-aza-benzimidazole | |
| N1C=NC2=CC=NC=C12 | |
| 119.13 | |
| 119.13 | |
| 5-Azabenzimidazole |
| 170°C to 173°C | |
| MFCD00051808 | |
| 2601 | |
| UBOOKRVGOBKDMM-UHFFFAOYSA-N | |
| 3H-imidazo[4,5-c]pyridine | |
| 9227 | |
| 98% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only