missing translation for 'onlineSavingsMsg'
Learn More

5-Azabenzimidazole, 98%, Thermo Scientific Chemicals

Catalog No. AAL0693003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0693003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0693003 Supplier Thermo Scientific Chemicals Supplier No. L0693003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

5-Azabenzimidazole is used as Pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Azabenzimidazole is used as Pharmaceutical intermediates.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.

Chemical Identifiers

CAS 272-97-9
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.13
MDL Number MFCD00051808
InChI Key UBOOKRVGOBKDMM-UHFFFAOYSA-N
Synonym 3h-imidazo 4,5-c pyridine, 1h-imidazo 4,5-c pyridine, 5-azabenzimidazole, 3,5-diazaindole, 5h-imidazo 4,5-c pyridine, imidazo 4,5-c pyridine, 5-aza-1h-benzoimidazole, 5-aza-benzimidazole
PubChem CID 9227
IUPAC Name 3H-imidazo[4,5-c]pyridine
SMILES N1C=NC2=CC=NC=C12

Specifications

Melting Point 170°C to 173°C
Quantity 1 g
Beilstein 2601
Formula Weight 119.13
Percent Purity 98%
Chemical Name or Material 5-Azabenzimidazole
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only