Learn More
5-Aza-2'-deoxycytidine, 98%
CAS: 2353-33-5 | C9H13N3O4 | 227.22 g/mol
Supplier: Thermo Scientific Chemicals J65513MC
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 5-Aza-2'-deoxycytidine | |
| White | |
| MFCD00006547 | |
| 617982 | |
| decitabine, 5-aza-2'-deoxycytidine, dacogen, 5-azadeoxycytidine, 2'-deoxy-5-azacytidine, azadc, dezocitidine, 5-aza-cdr, 5-aza-dc, dacogen tn | |
| NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1 | |
| 227.22 | |
| CHEBI:50131 | |
| 98% |
| 2353-33-5 | |
| C9H13N3O4 | |
| 100 mg | |
| 14,2853 | |
| CKTSBUTUHBMZGZ-SHYZEUOFSA-N | |
| 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,3,5-triazin-2-one | |
| 451668 | |
| 228.21 | |
| Powder |
Chemical Identifiers
| 2353-33-5 | |
| 227.22 | |
| CKTSBUTUHBMZGZ-SHYZEUOFSA-N | |
| 451668 | |
| 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,3,5-triazin-2-one |
| C9H13N3O4 | |
| MFCD00006547 | |
| decitabine, 5-aza-2'-deoxycytidine, dacogen, 5-azadeoxycytidine, 2'-deoxy-5-azacytidine, azadc, dezocitidine, 5-aza-cdr, 5-aza-dc, dacogen tn | |
| CHEBI:50131 | |
| NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1 |
Safety and Handling
GHS H Statement
H360-H341-H302-H315-H319-H335
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
H302-H315-H319-H335-H341-H360
EINECSNumber : 219-089-4
RTECSNumber : XZ3012000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only