missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Aminovaleric Acid 98.0+%, TCI America™
$115.57 - $363.49
Chemical Identifiers
| CAS | 660-88-8 |
|---|---|
| Molecular Formula | C5H11NO2 |
| Molecular Weight (g/mol) | 117.148 |
| MDL Number | MFCD00008232 |
| InChI Key | JJMDCOVWQOJGCB-UHFFFAOYSA-N |
| Synonym | 5-aminovaleric acid, 5-amino-pentanoic acid, homopiperidinic acid, 5-aminopentanoate, valeric acid, 5-amino, delta-amino-n-valeric acid, pentanoic acid, 5-amino, delta-aminovaleric acid, 5-amino-n-valeric acid, 5-aminovalerate |
| PubChem CID | 138 |
| ChEBI | CHEBI:15887 |
| IUPAC Name | 5-aminopentanoic acid |
| SMILES | C(CCN)CC(=O)O |
Chemical Identifiers
| 660-88-8 | |
| 117.148 | |
| JJMDCOVWQOJGCB-UHFFFAOYSA-N | |
| 138 | |
| 5-aminopentanoic acid |
| C5H11NO2 | |
| MFCD00008232 | |
| 5-aminovaleric acid, 5-amino-pentanoic acid, homopiperidinic acid, 5-aminopentanoate, valeric acid, 5-amino, delta-amino-n-valeric acid, pentanoic acid, 5-amino, delta-aminovaleric acid, 5-amino-n-valeric acid, 5-aminovalerate | |
| CHEBI:15887 | |
| C(CCN)CC(=O)O |
Specifications
| 158°C | |
| White-Yellow | |
| MFCD00008232 | |
| JJMDCOVWQOJGCB-UHFFFAOYSA-N | |
| 5-aminopentanoic acid | |
| 117.148 | |
| CHEBI:15887 | |
| ≥98.0% (T) | |
| 5-Aminovaleric Acid |
| 660-88-8 | |
| C5H11NO2 | |
| 5-aminovaleric acid, 5-amino-pentanoic acid, homopiperidinic acid, 5-aminopentanoate, valeric acid, 5-amino, delta-amino-n-valeric acid, pentanoic acid, 5-amino, delta-aminovaleric acid, 5-amino-n-valeric acid, 5-aminovalerate | |
| C(CCN)CC(=O)O | |
| 5 g | |
| 138 | |
| 117.15 | |
| Crystalline Powder |
Safety and Handling
TSCA : No