missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Aminovaleric Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 158°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 117.15 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-Aminovaleric Acid |
Safety and Handling
| TSCA | No |