missing translation for 'onlineSavingsMsg'
Learn More

5-Aminovaleric Acid Hydroiodide (Low water content) 97.0+%, TCI America™

$146.31 - $515.95

Chemical Identifiers

CAS 1705581-28-7
Molecular Formula C5H12INO2
Molecular Weight (g/mol) 245.06
InChI Key QRCPJIVRDACIKP-UHFFFAOYSA-N
Synonym 5-Aminopentanoic Acid Hydroiodide, Homopiperidinic Acid Hydroiodide, 5-AVAI
PubChem CID 118324268
IUPAC Name 5-aminopentanoic acid;hydroiodide
SMILES C(CCN)CC(=O)O.I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A29841G
View Documents
TCI America
A29841G
1 g
Each for $146.31
Only null left
 
A29845G
View Documents
TCI America
A29845G
5 g
Each for $515.95
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1705581-28-7
245.06
5-Aminopentanoic Acid Hydroiodide, Homopiperidinic Acid Hydroiodide, 5-AVAI
5-aminopentanoic acid;hydroiodide
C5H12INO2
QRCPJIVRDACIKP-UHFFFAOYSA-N
118324268
C(CCN)CC(=O)O.I

Specifications

1705581-28-7
C5H12INO2
5-Aminopentanoic Acid Hydroiodide, Homopiperidinic Acid Hydroiodide, 5-AVAI
C(CCN)CC(=O)O.I
245.06
245.06
Crystalline Powder
White-Yellow
1 g
QRCPJIVRDACIKP-UHFFFAOYSA-N
5-aminopentanoic acid;hydroiodide
118324268
≥97.0% (N,T)
5-Aminovaleric Acid Hydroiodide (Low water content)
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents