missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Aminovaleric Acid Hydroiodide (Low water content) 97.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 245.06 |
| Percent Purity | ≥97.0% (N,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-Aminovaleric Acid Hydroiodide (Low water content) |
Safety and Handling
| TSCA | No |