missing translation for 'onlineSavingsMsg'
Learn More

5-Aminovaleric Acid Hydrochloride (Low water content) 98.0+%, TCI America™

Catalog No. p-7116900
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A04361G 1 g
A04365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A04361G Supplier TCI America Supplier No. A04361G
Only null left

Chemical Identifiers

CAS 627-95-2
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.606
MDL Number MFCD00012919
InChI Key BLOIUFYKQCCAGP-UHFFFAOYSA-N
Synonym 5-aminovaleric acid hydrochloride, 5-aminopentanoic acid hydrochloride, 4-carboxybutylammonium chloride, 5-aminovaleric acid hcl, 5-amino-n-valeric acid hydrochloride, pentanoic acid, 5-amino-, hydrochloride, 4-carboxybutan-1-aminium chloride, acmc-1az5k, spectrum1501126, homopiperidinic acid hydrochloride
PubChem CID 69398
IUPAC Name 5-aminopentanoic acid;hydrochloride
SMILES C(CCN)CC(=O)O.Cl

Specifications

Melting Point 96°C
Quantity 1 g
Formula Weight 153.61
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminovaleric Acid Hydrochloride (Low water content)