missing translation for 'onlineSavingsMsg'
Learn More

5-Aminovaleric acid, 97%

Catalog No. AC153910250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC153910250 25 g
AC153910050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC153910250 Supplier Thermo Scientific Chemicals Supplier No. 153910250
Only null left

CAS: 660-88-8 | C5H11NO2 | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 660-88-8
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00008232
InChI Key JJMDCOVWQOJGCB-UHFFFAOYSA-N
Synonym 5-aminovaleric acid, 5-amino-pentanoic acid, homopiperidinic acid, 5-aminopentanoate, valeric acid, 5-amino, delta-amino-n-valeric acid, pentanoic acid, 5-amino, delta-aminovaleric acid, 5-amino-n-valeric acid, 5-aminovalerate
PubChem CID 138
ChEBI CHEBI:15887
IUPAC Name 5-aminopentanoic acid
SMILES C(CCN)CC(=O)O

Specifications

Melting Point 158.0°C to 161.0°C
CAS Min % 96.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula H2N(CH2)4CO2H
Beilstein 04, 418
Solubility Information solution
Quantity 25 g
Formula Weight 117.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminovaleric acid, 97%
Show More Show Less
EINECSNumber 211-544-5

RUO – Research Use Only