missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Aminouracil 98.0+%, TCI America™
Supplier: TCI America A089825G
Specifications
| 5-Aminouracil | |
| C4H5N3O2 | |
| 25 g | |
| BISHACNKZIBDFM-UHFFFAOYSA-N | |
| 5-amino-1,2,3,4-tetrahydropyrimidine-2,4-dione | |
| 13611 | |
| 127.10 | |
| Crystalline Powder |
| 932-52-5 | |
| MFCD00006025 | |
| 5-aminouracil, 5-amino-2,4-dihydroxypyrimidine, uracil, 5-amino, 2,4 1h,3h-pyrimidinedione, 5-amino, 2,4-dihydroxy-5-aminopyrimidine, unii-r2o7eky9g9, r2o7eky9g9, 5-aminopyrimidine-2,4-diol, 5-amino-1,3-dihydropyrimidine-2,4-dione, wbu | |
| NC1=CNC(=O)NC1=O | |
| 127.10 | |
| CHEBI:46348 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 932-52-5 | |
| 127.10 | |
| BISHACNKZIBDFM-UHFFFAOYSA-N | |
| 13611 | |
| 5-amino-1,2,3,4-tetrahydropyrimidine-2,4-dione |
| C4H5N3O2 | |
| MFCD00006025 | |
| 5-aminouracil, 5-amino-2,4-dihydroxypyrimidine, uracil, 5-amino, 2,4 1h,3h-pyrimidinedione, 5-amino, 2,4-dihydroxy-5-aminopyrimidine, unii-r2o7eky9g9, r2o7eky9g9, 5-aminopyrimidine-2,4-diol, 5-amino-1,3-dihydropyrimidine-2,4-dione, wbu | |
| CHEBI:46348 | |
| NC1=CNC(=O)NC1=O |
Safety and Handling
RTECSNumber : YQ8740000
TSCA : Yes