missing translation for 'onlineSavingsMsg'
Learn More

5-Aminouracil 98.0+%, TCI America™

Catalog No. A08985G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A08985G 5 g
A089825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A08985G Supplier TCI America Supplier No. A08985G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 932-52-5
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.10
MDL Number MFCD00006025
InChI Key BISHACNKZIBDFM-UHFFFAOYSA-N
Synonym 5-aminouracil, 5-amino-2,4-dihydroxypyrimidine, uracil, 5-amino, 2,4 1h,3h-pyrimidinedione, 5-amino, 2,4-dihydroxy-5-aminopyrimidine, unii-r2o7eky9g9, r2o7eky9g9, 5-aminopyrimidine-2,4-diol, 5-amino-1,3-dihydropyrimidine-2,4-dione, wbu
PubChem CID 13611
ChEBI CHEBI:46348
IUPAC Name 5-amino-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES NC1=CNC(=O)NC1=O

Specifications

Quantity 5 g
Formula Weight 127.10
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminouracil
RTECSNumber YQ8740000
TSCA Yes