missing translation for 'onlineSavingsMsg'
Learn More

5-Aminouracil, 98%, Thermo Scientific™

$255.92

Chemical Identifiers

CAS 932-52-5
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.10
MDL Number MFCD00006025
InChI Key BISHACNKZIBDFM-UHFFFAOYSA-N
Synonym 5-aminouracil, 5-amino-2,4-dihydroxypyrimidine, uracil, 5-amino, 2,4 1h,3h-pyrimidinedione, 5-amino, 2,4-dihydroxy-5-aminopyrimidine, unii-r2o7eky9g9, r2o7eky9g9, 5-aminopyrimidine-2,4-diol, 5-amino-1,3-dihydropyrimidine-2,4-dione, wbu
PubChem CID 13611
ChEBI CHEBI:46348
IUPAC Name 5-amino-1H-pyrimidine-2,4-dione
SMILES NC1=CNC(=O)NC1=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC226820250
View Documents Promotion Details
N/A 25g Glass bottle
Each for $255.92
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

932-52-5
127.10
BISHACNKZIBDFM-UHFFFAOYSA-N
13611
5-amino-1H-pyrimidine-2,4-dione
C4H5N3O2
MFCD00006025
5-aminouracil, 5-amino-2,4-dihydroxypyrimidine, uracil, 5-amino, 2,4 1h,3h-pyrimidinedione, 5-amino, 2,4-dihydroxy-5-aminopyrimidine, unii-r2o7eky9g9, r2o7eky9g9, 5-aminopyrimidine-2,4-diol, 5-amino-1,3-dihydropyrimidine-2,4-dione, wbu
CHEBI:46348
NC1=CNC(=O)NC1=O

Specifications

932-52-5
100.0
97.5% min. (HPLC)
MFCD00006025
5-aminouracil, 5-amino-2,4-dihydroxypyrimidine, uracil, 5-amino, 2,4 1h,3h-pyrimidinedione, 5-amino, 2,4-dihydroxy-5-aminopyrimidine, unii-r2o7eky9g9, r2o7eky9g9, 5-aminopyrimidine-2,4-diol, 5-amino-1,3-dihydropyrimidine-2,4-dione, wbu
BISHACNKZIBDFM-UHFFFAOYSA-N
5-amino-1H-pyrimidine-2,4-dione
13611
127.1
5-Aminouracil, 98%
97.5
Authentic
C4H5N3O2
24, 463
Solubility in water: 0.5g/L (20°C)
NC1=CNC(=O)NC1=O
127.10
CHEBI:46348
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Warning

EINECSNumber : 213-252-3

RTECSNumber : YQ8740000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only