missing translation for 'onlineSavingsMsg'
Learn More

5-Aminoquinoline

Catalog No. AC104600010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104600010 1 g Glass bottle
AC104600050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC104600010 Supplier Thermo Scientific Chemicals Supplier No. 104600010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 611-34-7 | C9H8N2 | 144.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 611-34-7
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00006797
InChI Key XMIAFAKRAAMSGX-UHFFFAOYSA-N
Synonym 5-aminoquinoline, 5-quinolinamine, quinoline, 5-amino, 5-quinolylamine, quinolin-5-ylamine, unii-yyj34z13sh, 5-amino-quinoline, ccris 1680, yyj34z13sh
PubChem CID 11911
IUPAC Name quinolin-5-amine
SMILES NC1=C2C=CC=NC2=CC=C1

Specifications

Melting Point 106.0°C to 109.0°C
Color White to Yellow
Boiling Point 310.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1 g
Beilstein 22, 445
Formula Weight 144.18
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder or Granules
Chemical Name or Material 5-Aminoquinoline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 210-266-1

RUO – Research Use Only