missing translation for 'onlineSavingsMsg'
Learn More

5-Aminoquinoline 99.0+%, TCI America™

Catalog No. p-7117157
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A09595G 5 g
A095925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A09595G Supplier TCI America Supplier No. A09595G
Only null left

Chemical Identifiers

CAS 611-34-7
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00006797
InChI Key XMIAFAKRAAMSGX-UHFFFAOYSA-N
Synonym 5-aminoquinoline, 5-quinolinamine, quinoline, 5-amino, 5-quinolylamine, quinolin-5-ylamine, unii-yyj34z13sh, 5-amino-quinoline, ccris 1680, yyj34z13sh
PubChem CID 11911
IUPAC Name quinolin-5-amine
SMILES NC1=C2C=CC=NC2=CC=C1

Specifications

Melting Point 109°C
Boiling Point 310°C
Quantity 5 g
Formula Weight 144.18
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminoquinoline
RTECSNumber VA9625000
TSCA No