missing translation for 'onlineSavingsMsg'
Learn More

5-Aminophthalide, 97%, Thermo Scientific Chemicals

Catalog No. p-7057104
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6645703 1 g
AAH6645706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6645703 Supplier Thermo Scientific Chemicals Supplier No. H6645703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 65399-05-5 | C8H7NO2 | 149.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65399-05-5
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.149
MDL Number MFCD00778315
InChI Key ISMUWQMUWFPFBZ-UHFFFAOYSA-N
Synonym 5-aminophthalide, 5-aminoisobenzofuran-1 3h-one, 5-amino-3h-isobenzofuran-1-one, 5-amino-2-benzofuran-1 3h-one, 1 3h-isobenzofuranone, 5-amino, 5-amino-1 3h-isobenzofuranone, 5-amino-1,3-dihydro-2-benzofuran-1-one, 5-amino-3-hydroisobenzofuran-1-one, 5-aminophthliade, 5-amino-phthalide
PubChem CID 720669
IUPAC Name 5-amino-3H-2-benzofuran-1-one
SMILES C1C2=C(C=CC(=C2)N)C(=O)O1

Specifications

Quantity 1 g
Beilstein 121983
Formula Weight 149.05
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Aminophthalide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 265-731-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only