missing translation for 'onlineSavingsMsg'
Learn More

5-Aminomethyl-2-(Boc-amino)pyridine, 97%

Catalog No. AAH5007803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5007803 1 g
AAH5007806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5007803 Supplier Thermo Scientific Chemicals Supplier No. H5007803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 187237-37-2 | C11H17N3O2 | 223.276 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 187237-37-2
Molecular Formula C11H17N3O2
Molecular Weight (g/mol) 223.276
MDL Number MFCD07781132
InChI Key DFLQTVPEIMTXSZ-UHFFFAOYSA-N
Synonym 2-boc-amino-5-aminomethyl pyridine, 5-aminomethyl-2-n-boc aminopyridine, tert-butyl 5-aminomethyl pyridin-2-yl carbamate, tert-butyl 5-aminomethyl pyridin-2-ylcarbamate, tert-butyl n-5-aminomethyl pyridin-2-yl carbamate, 5-aminomethyl-pyridin-2-yl-carbamic acid tert-butyl ester, 5-aminomethyl-2-boc-amino pyridine, carbamic acid,n-5-aminomethyl-2-pyridinyl-, 1,1-dimethylethyl ester, 2-tert-butoxycarbonyl-amino-5-aminomethylpyridine
PubChem CID 42553118
IUPAC Name tert-butyl N-[5-(aminomethyl)pyridin-2-yl]carbamate
SMILES CC(C)(C)OC(=O)NC1=NC=C(C=C1)CN

Specifications

Melting Point 132°C to 134°C
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 223.28
Percent Purity 97%
Chemical Name or Material 5-Aminomethyl-2-(Boc-amino)pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only