missing translation for 'onlineSavingsMsg'
Learn More

5-Aminolevulinic Acid Hydrochloride 98.0+%, TCI America™

Catalog No. A03251G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A03251G 1 g
A0325100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A03251G Supplier TCI America Supplier No. A03251G

Chemical Identifiers

CAS 9-2-5451
Molecular Formula C5H10ClNO3
Molecular Weight (g/mol) 167.59
MDL Number MFCD00012869
InChI Key ZLHFONARZHCSET-UHFFFAOYSA-N
Synonym 5-aminolevulinic acid hydrochloride, 5-amino-4-oxopentanoic acid hydrochloride, aminolevulinic acid hydrochloride, levulan, levulan kerastick, aminolevulinic acid hcl, delta-aminolevulinic acid hydrochloride, gliolan, ameluz, 5-aminolevulinate hydrochloride
PubChem CID 123608
IUPAC Name 5-amino-4-oxopentanoic acid;hydrochloride
SMILES C(CC(=O)O)C(=O)CN.Cl

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 167.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminolevulinic Acid Hydrochloride
RTECSNumber OI1640000
TSCA No