missing translation for 'onlineSavingsMsg'
Learn More

5-Aminoindole 98.0+%, TCI America™

Catalog No. A20711G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20711G 1 g
A20715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20711G Supplier TCI America Supplier No. A20711G

Chemical Identifiers

CAS 5192-03-0
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.17
MDL Number MFCD00005679
InChI Key ZCBIFHNDZBSCEP-UHFFFAOYSA-N
Synonym 5-aminoindole, indol-5-ylamine, indole, 5-amino, unii-q732pg0y16, 1h-indol-5-ylamine, indole-5-ylamine, 5-indolamine, 5-amino indole, 5-amino-indole
PubChem CID 78867
ChEBI CHEBI:33067
IUPAC Name 1H-indol-5-amine
SMILES NC1=CC=C2NC=CC2=C1

Specifications

Melting Point 133°C
Boiling Point 190°C
Quantity 1 g
Formula Weight 132.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminoindole