missing translation for 'onlineSavingsMsg'
Learn More

5-Aminoindole, 97%

Catalog No. AC103900010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC103900010 1 g Glass bottle
AC103900050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC103900010 Supplier Thermo Scientific Chemicals Supplier No. 103900010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5192-03-0 | C8H8N2 | 132.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5192-03-0
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.17
MDL Number MFCD00005679
InChI Key ZCBIFHNDZBSCEP-UHFFFAOYSA-N
Synonym 5-aminoindole, indol-5-ylamine, indole, 5-amino, unii-q732pg0y16, 1h-indol-5-ylamine, indole-5-ylamine, 5-indolamine, 5-amino indole, 5-amino-indole
PubChem CID 78867
ChEBI CHEBI:33067
IUPAC Name 1H-indol-5-amine
SMILES NC1=CC=C2NC=CC2=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 130.0°C to 134.0°C
Color Brown-Gray to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in most common organic solvents
Formula Weight 132.16
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminoindole, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent
EINECSNumber 225-977-2
TSCA TSCA
Recommended Storage −20°C

RUO – Research Use Only