missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Aminoindazole 98.0+%, TCI America™
$134.75 - $483.21
Chemical Identifiers
| CAS | 19335-11-6 |
|---|---|
| Molecular Formula | C7H7N3 |
| Molecular Weight (g/mol) | 133.154 |
| MDL Number | MFCD00037975 |
| InChI Key | XBTOSRUBOXQWBO-UHFFFAOYSA-N |
| Synonym | 5-aminoindazole, 5-amino-1h-indazole, 2h-indazol-5-amine, 5-indazolamine, 1h-indazole-5-amine, 1h-indazol-5-ylamine, indazol-5-ylamine, 1h-indazole, 5-amino, 5-aminobenzopyrazole, 5-amino-indazole |
| PubChem CID | 88012 |
| IUPAC Name | 1H-indazol-5-amine |
| SMILES | C1=CC2=C(C=C1N)C=NN2 |
Chemical Identifiers
| 19335-11-6 | |
| 133.154 | |
| XBTOSRUBOXQWBO-UHFFFAOYSA-N | |
| 88012 | |
| C1=CC2=C(C=C1N)C=NN2 |
| C7H7N3 | |
| MFCD00037975 | |
| 5-aminoindazole, 5-amino-1h-indazole, 2h-indazol-5-amine, 5-indazolamine, 1h-indazole-5-amine, 1h-indazol-5-ylamine, indazol-5-ylamine, 1h-indazole, 5-amino, 5-aminobenzopyrazole, 5-amino-indazole | |
| 1H-indazol-5-amine |
Specifications
| 19335-11-6 | |
| C7H7N3 | |
| 5 g | |
| 5-aminoindazole, 5-amino-1h-indazole, 2h-indazol-5-amine, 5-indazolamine, 1h-indazole-5-amine, 1h-indazol-5-ylamine, indazol-5-ylamine, 1h-indazole, 5-amino, 5-aminobenzopyrazole, 5-amino-indazole | |
| C1=CC2=C(C=C1N)C=NN2 | |
| 133.154 | |
| 133.15 | |
| Crystalline Powder |
| 177°C | |
| MFCD00037975 | |
| 2811 | |
| XBTOSRUBOXQWBO-UHFFFAOYSA-N | |
| 1H-indazol-5-amine | |
| 88012 | |
| ≥98.0% (HPLC,T) | |
| 5-Aminoindazole |
Safety and Handling
RTECSNumber : NK7711000
TSCA : Yes