missing translation for 'onlineSavingsMsg'
Learn More

5-Aminoindazole 98.0+%, TCI America™

Catalog No. A07975G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A07975G 5 g
A079725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A07975G Supplier TCI America Supplier No. A07975G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19335-11-6
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.154
MDL Number MFCD00037975
InChI Key XBTOSRUBOXQWBO-UHFFFAOYSA-N
Synonym 5-aminoindazole, 5-amino-1h-indazole, 2h-indazol-5-amine, 5-indazolamine, 1h-indazole-5-amine, 1h-indazol-5-ylamine, indazol-5-ylamine, 1h-indazole, 5-amino, 5-aminobenzopyrazole, 5-amino-indazole
PubChem CID 88012
IUPAC Name 1H-indazol-5-amine
SMILES C1=CC2=C(C=C1N)C=NN2

Specifications

Melting Point 177°C
Quantity 5 g
UN Number 2811
Formula Weight 133.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminoindazole
RTECSNumber NK7711000
TSCA Yes