missing translation for 'onlineSavingsMsg'
Learn More

5-Aminoimidazole-4-carboxamide, 95%

Catalog No. AC397550250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397550250 25 g Glass bottle
AC397550050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC397550250 Supplier Thermo Scientific Chemicals Supplier No. 397550250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 360-97-4 | C4H6N4O | 126.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 360-97-4
Molecular Formula C4H6N4O
Molecular Weight (g/mol) 126.12
MDL Number MFCD02181040
InChI Key DVNYTAVYBRSTGK-UHFFFAOYSA-N
Synonym 5-aminoimidazole-4-carboxamide, 5-amino-1h-imidazole-4-carboxamide, 4-amino-5-imidazolecarboxamide, 5-amino-4-imidazolecarboxamide, colahepat, 4-aminoimidazole-5-carboxamide, aica, 1h-imidazole-4-carboxamide, 5-amino, diazol-c, 4-carbamoyl-5-aminoimidazole
PubChem CID 9679
ChEBI CHEBI:2030
IUPAC Name 4-amino-1H-imidazole-5-carboxamide
SMILES NC(=O)C1=C(N)N=CN1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 164.0°C to 170.0°C
Color Gray to Purple
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 126.12
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminoimidazole-4-carboxamide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-641-4

RUO – Research Use Only