missing translation for 'onlineSavingsMsg'
Learn More

5-Aminoimidazole-4-carboxamide-1-beta-D-ribofuranoside, 98%

Catalog No. AC398921000
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC398921000 100 mg Glass bottle
AC398920050 5 mg Glass bottle
AC398920250 25 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC398921000 Supplier Thermo Scientific Chemicals Supplier No. 398921000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2627-69-2 | C9H14N4O5 | 258.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2627-69-2
Molecular Formula C9H14N4O5
Molecular Weight (g/mol) 258.23
MDL Number MFCD00869751
InChI Key RTRQQBHATOEIAF-UUOKFMHZSA-N
Synonym acadesine, aica-riboside, aicar, arasine, aica riboside, aic-riboside, acadesina, acadesinum, 5-amino-1-beta-d-ribofuranosyl-1h-imidazole-4-carboxamide, protara
PubChem CID 17513
ChEBI CHEBI:28498
IUPAC Name 5-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]imidazole-4-carboxamide
SMILES NC(=O)C1=C(N)N(C=N1)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 214°C to 216°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 mg
Merck Index 15, 17
Specific Rotation -62.0° to -66.0° (20°C, 589nm) (c=1 H2O)
Solubility Information Solubility in water: soluble (7g/L at 25°C). Other solubilities: solubility in aq. pbs (2.5g/L at 25°C),soluble in ethanol,dmso
Formula Weight 258.23
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranoside
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 220-097-5

RUO – Research Use Only