missing translation for 'onlineSavingsMsg'
Learn More

5-Aminofluorescein (isomer I) 95.0+%, TCI America™

Catalog No. p-7116822
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A03061G 1 g
A03065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A03061G Supplier TCI America Supplier No. A03061G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3326-34-9
Molecular Formula C20H13NO5
Molecular Weight (g/mol) 347.33
MDL Number MFCD00005052
InChI Key GZAJOEGTZDUSKS-UHFFFAOYSA-N
Synonym 5-aminofluorescein, fluoresceinamine, 4-aminofluorescein, fluoresceinamine isomer i, fluoresceinamine, isomer i, 1-aminofluorescein, spiro isobenzofuran-1 3h ,9'-9h xanthen-3-one, 5-amino-3',6'-dihydroxy, 5 6-aminofluorescein mixture of isomers, 5-amino-3',6'-dihydroxyspiro 2-benzofuran-1,9'-xanthene-3-one, 5-amino-3',6'-dihydroxy-3h-spiro 2-benzofuran-1,9'-xanthene-3-one
PubChem CID 76845
IUPAC Name 5-amino-3',6'-dihydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
SMILES NC1=CC=C2C(=C1)C(=O)OC21C2=CC=C(O)C=C2OC2=CC(O)=CC=C12

Specifications

Quantity 1 g
Formula Weight 347.33
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminofluorescein (isomer I)