Learn More
5-Aminobenzothiazole, 95%, Thermo Scientific™
Chemical Identifiers
| CAS | 1123-93-9 |
|---|---|
| Molecular Formula | C7H6N2S |
| Molecular Weight (g/mol) | 150.20 |
| MDL Number | MFCD04115282 |
| InChI Key | UJZYHMZRXGNDFB-UHFFFAOYSA-N |
| Synonym | 5-benzothiazolamine, benzothiazol-5-ylamine, benzo d thiazol-5-amine, benzothiazol-5-amine, 5-amino-1,3-benzothiazole, 5-aminobenzothiazole, 1,3-benzothiazole-5-amine, 1,3-benzothiazol-5-ylamine, 5-amino-1,3-benzothiazol, 5-amino-benzothiazole |
| PubChem CID | 70749 |
| IUPAC Name | 1,3-benzothiazol-5-amine |
| SMILES | NC1=CC=C2SC=NC2=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC451540050
|
Thermo Scientific Chemicals
451540050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC451540250
|
Thermo Scientific Chemicals
451540250 |
25g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 1123-93-9 | |
| 150.20 | |
| UJZYHMZRXGNDFB-UHFFFAOYSA-N | |
| 70749 | |
| NC1=CC=C2SC=NC2=C1 |
| C7H6N2S | |
| MFCD04115282 | |
| 5-benzothiazolamine, benzothiazol-5-ylamine, benzo d thiazol-5-amine, benzothiazol-5-amine, 5-amino-1,3-benzothiazole, 5-aminobenzothiazole, 1,3-benzothiazole-5-amine, 1,3-benzothiazol-5-ylamine, 5-amino-1,3-benzothiazol, 5-amino-benzothiazole | |
| 1,3-benzothiazol-5-amine |
Specifications
| 1123-93-9 | |
| 94% min. (HPLC) | |
| MFCD04115282 | |
| 5-benzothiazolamine, benzothiazol-5-ylamine, benzo d thiazol-5-amine, benzothiazol-5-amine, 5-amino-1,3-benzothiazole, 5-aminobenzothiazole, 1,3-benzothiazole-5-amine, 1,3-benzothiazol-5-ylamine, 5-amino-1,3-benzothiazol, 5-amino-benzothiazole | |
| NC1=CC=C2SC=NC2=C1 | |
| 150.20 | |
| 150.2 | |
| 5-Aminobenzothiazole |
| Authentic | |
| C7H6N2S | |
| 5g | |
| UJZYHMZRXGNDFB-UHFFFAOYSA-N | |
| 1,3-benzothiazol-5-amine | |
| 70749 | |
| 95% |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
EINECSNumber : 214-381-8
RUO â Research Use Only