missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Aminobenzothiazole, 95%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 73.0°C to 77.5°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 94% min. (HPLC) |
| Quantity | 25 g |
| Formula Weight | 150.2 |
| Percent Purity | 95% |
| Physical Form | Powder or Solid |
| Chemical Name or Material | 5-Aminobenzothiazole |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H StatementHarmful in contact with skin.Causes skin irritation.May cause respiratory irritation.Causes serious eye irritation.Harmful if swallowed.Harmful if inhaled. |
| Precautionary Statement | GHS P StatementWear protective gloves/protective clothing/eye protection/face protection.IF ON SKIN: Wash with plenty of soap and water.IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.IF |
| EINECSNumber | 214-381-8 |
RUO – Research Use Only