missing translation for 'onlineSavingsMsg'
Learn More

5-Aminobenzothiazole, 95%

Catalog No. AC451540050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC451540050 5 g
AC451540250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC451540050 Supplier Thermo Scientific Chemicals Supplier No. 451540050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1123-93-9 | C7H6N2S | 150.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1123-93-9
Molecular Formula C7H6N2S
Molecular Weight (g/mol) 150.20
MDL Number MFCD04115282
InChI Key UJZYHMZRXGNDFB-UHFFFAOYSA-N
Synonym 5-benzothiazolamine, benzothiazol-5-ylamine, benzo d thiazol-5-amine, benzothiazol-5-amine, 5-amino-1,3-benzothiazole, 5-aminobenzothiazole, 1,3-benzothiazole-5-amine, 1,3-benzothiazol-5-ylamine, 5-amino-1,3-benzothiazol, 5-amino-benzothiazole
PubChem CID 70749
IUPAC Name 1,3-benzothiazol-5-amine
SMILES NC1=CC=C2SC=NC2=C1

Specifications

Melting Point 73.0°C to 77.5°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 5 g
Formula Weight 150.2
Percent Purity 95%
Chemical Name or Material 5-Aminobenzothiazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 214-381-8

RUO – Research Use Only