missing translation for 'onlineSavingsMsg'
Learn More

5-Aminobenzimidazole 98.0+%, TCI America™

Catalog No. A26051G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26051G 1 g
A26055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26051G Supplier TCI America Supplier No. A26051G

Chemical Identifiers

CAS 934-22-5
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.15
MDL Number MFCD00465258,MFCD00831692
InChI Key WFRXSXUDWCVSPI-UHFFFAOYSA-N
PubChem CID 13623
IUPAC Name 1H-1,3-benzodiazol-6-amine
SMILES NC1=CC=C2N=CNC2=C1

Specifications

Melting Point 163°C
Boiling Point 222°C
Quantity 1 g
Formula Weight 133.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminobenzimidazole
RTECSNumber DD5785000
TSCA No