missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-6-chloro-2,3-dicyanopyrazine 98.0+%, TCI America™

Catalog No. d226910g
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D226910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D226910G Supplier TCI America Supplier No. D226910G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56413-96-8
Molecular Formula C6H2ClN5
Molecular Weight (g/mol) 179.57
MDL Number MFCD00191417
InChI Key CDKQWBZSAPRZNS-UHFFFAOYSA-N
Synonym 5-Amino-6-chloro-2,3-pyrazinedicarbonitrile
PubChem CID 601645
IUPAC Name 5-amino-6-chloropyrazine-2,3-dicarbonitrile
SMILES NC1=NC(C#N)=C(N=C1Cl)C#N

Specifications

Melting Point 208°C
Quantity 10 g
UN Number 2811
Formula Weight 179.57
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-6-chloro-2,3-dicyanopyrazine