missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-4-chloropyrimidine 97.0+%, TCI America™

Catalog No. p-7118155
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2937200MG 200 mg
A29371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2937200MG Supplier TCI America Supplier No. A2937200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 54660-78-5
Molecular Formula C4H4ClN3
Molecular Weight (g/mol) 129.547
MDL Number MFCD06412561
InChI Key LHGMCUVJFRBVBH-UHFFFAOYSA-N
Synonym 5-amino-4-chloropyrimidine, 4-chloro-5-aminopyrimidine, 4-chloranylpyrimidin-5-amine, 5-pyrimidinamine, 4-chloro, 4-chloro-5-amino-pyrimidine, acmc-209lid, 4-chloro-5-pyrimidinamine, 5-amino-6-chloropyrimidine, 4-chloropyrimidine-5-amine, 4-chloropyrimidin-5-ylamine
PubChem CID 13404541
IUPAC Name 4-chloropyrimidin-5-amine
SMILES C1=C(C(=NC=N1)Cl)N

Specifications

Melting Point 108°C
Color White-Yellow
Quantity 200 mg
Formula Weight 129.55
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-4-chloropyrimidine
TSCA No
Recommended Storage Refrigerator