missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-4-chloropyrimidine, 96%

Catalog No. p-7057222
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6667103 1 g
AAH6667106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6667103 Supplier Thermo Scientific Chemicals Supplier No. H6667103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 54660-78-5 | C4H4ClN3 | 129.547 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54660-78-5
Molecular Formula C4H4ClN3
Molecular Weight (g/mol) 129.547
MDL Number MFCD06412561
InChI Key LHGMCUVJFRBVBH-UHFFFAOYSA-N
Synonym 5-amino-4-chloropyrimidine, 4-chloro-5-aminopyrimidine, 4-chloranylpyrimidin-5-amine, 5-pyrimidinamine, 4-chloro, 4-chloro-5-amino-pyrimidine, acmc-209lid, 4-chloro-5-pyrimidinamine, 5-amino-6-chloropyrimidine, 4-chloropyrimidine-5-amine, 4-chloropyrimidin-5-ylamine
PubChem CID 13404541
IUPAC Name 4-chloropyrimidin-5-amine
SMILES C1=C(C(=NC=N1)Cl)N

Specifications

Melting Point 110°C to 116°C
Color Brown to White
Quantity 1 g
Sensitivity Air and light sensitive
Formula Weight 129.55
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 5-Amino-4-chloropyrimidine
Hazard Category H302-H315-H318-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only