missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-4,6-dichloro-2-(propylthio)pyrimidine 98.0+%, TCI America™

Catalog No. A27165G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27165G 5 g
A27161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27165G Supplier TCI America Supplier No. A27165G
Only null left

Chemical Identifiers

CAS 145783-15-9
Molecular Formula C7H9Cl2N3S
Molecular Weight (g/mol) 238.13
MDL Number MFCD16875503
InChI Key CJJLJBFJNXMANZ-UHFFFAOYSA-N
PubChem CID 11436336
IUPAC Name 4,6-dichloro-2-(propylsulfanyl)pyrimidin-5-amine
SMILES CCCSC1=NC(Cl)=C(N)C(Cl)=N1

Specifications

Quantity 5 g
Formula Weight 238.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-4,6-dichloro-2-(propylthio)pyrimidine