missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-4,6-dichloro-2-methylpyrimidine 98.0+%, TCI America™

Catalog No. p-7117647
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19465G 5 g
A194625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19465G Supplier TCI America Supplier No. A19465G
Only null left

Chemical Identifiers

CAS 39906-04-2
Molecular Formula C5H5Cl2N3
Molecular Weight (g/mol) 178.016
MDL Number MFCD00194053
InChI Key FKRXXAMAHOGYNT-UHFFFAOYSA-N
Synonym 5-amino-4,6-dichloro-2-methylpyrimidine, 2-methyl-4,6-dichloro-5-aminopyrimidine, 4,6-dichloro-2-methyl-pyrimidin-5-ylamine, 5-pyrimidinamine, 4,6-dichloro-2-methyl, 4,6-dichloro-2-methyl-pyrimidin-5-amine, 4,6-dichloro-2-methylpyrimidin-5-ylamine, pubchem6960, acmc-209j8x, ksc496o2d, fkrxxamahogynt-uhfffaoysa
PubChem CID 736674
IUPAC Name 4,6-dichloro-2-methylpyrimidin-5-amine
SMILES CC1=NC(=C(C(=N1)Cl)N)Cl

Specifications

Melting Point 73°C
Color Yellow
Quantity 5 g
Formula Weight 178.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-4,6-dichloro-2-methylpyrimidine