missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-3-tert-butyl-1-methylpyrazole, 98%

Catalog No. AAB2009503
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2009503 1 g
AAB2009506 5 g
AAB2009514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2009503 Supplier Thermo Scientific Chemicals Supplier No. B2009503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 118430-73-2 | C8H15N3 | 153.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 118430-73-2
Molecular Formula C8H15N3
Molecular Weight (g/mol) 153.23
MDL Number MFCD00068002
InChI Key XSCDSAMVQLKDNI-UHFFFAOYSA-N
Synonym 5-amino-3-tert-butyl-1-methylpyrazole, 3-tert-butyl-1-methyl-1h-pyrazol-5-amine, 5-tert-butyl-2-methyl-2h-pyrazol-3-ylamine, 1h-pyrazol-5-amine, 3-1,1-dimethylethyl-1-methyl, 5-amino-3-tert-butyl-1-methyl-1h-pyrazole, 3-tert-butyl-1-methylpyrazole-5-ylamine, acmc-1cins, maybridge4_002215, 5-amino-1-methyl-3-tert-butylpyrazole
PubChem CID 2735287
SMILES CN1N=C(C=C1N)C(C)(C)C

Specifications

Melting Point 155°C to 158°C
Quantity 1 g
IUPAC Name 3-tert-butyl-1-methyl-1H-pyrazol-5-amine
Formula Weight 153.22
Percent Purity 98%
Chemical Name or Material 5-Amino-3-tert-butyl-1-methylpyrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only