missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-3-phenylisoxazole 98.0+%, TCI America™

$165.90 - $605.83

Chemical Identifiers

CAS 4369-55-5
Molecular Formula C9H8N2O
Molecular Weight (g/mol) 160.18
MDL Number MFCD00464114
InChI Key HLOHVVZZMMMDMM-UHFFFAOYSA-N
Synonym 3-phenylisoxazol-5-amine, 5-amino-3-phenylisoxazole, 5-isoxazolamine, 3-phenyl, 3-phenyl-5-isoxazolamine, 3-phenylisoxazole-5-ylamine, isoxazole, 5-amino-3-phenyl, phenylisoxazolamine, pubchem15538, 3-phenylisoxazole-5-amine, acmc-209jv8
PubChem CID 261201
IUPAC Name 3-phenyl-1,2-oxazol-5-amine
SMILES NC1=CC(=NO1)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A24371G
View Documents
TCI America
A24371G
1 g
Each for $165.90
Only null left
 
A24375G
View Documents
TCI America
A24375G
5 g
Each for $605.83
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4369-55-5
160.18
HLOHVVZZMMMDMM-UHFFFAOYSA-N
261201
NC1=CC(=NO1)C1=CC=CC=C1
C9H8N2O
MFCD00464114
3-phenylisoxazol-5-amine, 5-amino-3-phenylisoxazole, 5-isoxazolamine, 3-phenyl, 3-phenyl-5-isoxazolamine, 3-phenylisoxazole-5-ylamine, isoxazole, 5-amino-3-phenyl, phenylisoxazolamine, pubchem15538, 3-phenylisoxazole-5-amine, acmc-209jv8
3-phenyl-1,2-oxazol-5-amine

Specifications

4369-55-5
White
MFCD00464114
3-phenylisoxazol-5-amine, 5-amino-3-phenylisoxazole, 5-isoxazolamine, 3-phenyl, 3-phenyl-5-isoxazolamine, 3-phenylisoxazole-5-ylamine, isoxazole, 5-amino-3-phenyl, phenylisoxazolamine, pubchem15538, 3-phenylisoxazole-5-amine, acmc-209jv8
NC1=CC(=NO1)C1=CC=CC=C1
160.18
160.18
Crystalline Powder
113°C
C9H8N2O
1 g
HLOHVVZZMMMDMM-UHFFFAOYSA-N
3-phenyl-1,2-oxazol-5-amine
261201
≥98.0% (HPLC,N)
5-Amino-3-phenylisoxazole
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'