missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-3-(4-methoxyphenyl)-1H-pyrazole, 97%

Catalog No. AAH3158003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3158003 1 g
AAH3158006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3158003 Supplier Thermo Scientific Chemicals Supplier No. H3158003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 19541-95-8 | C10H11N3O | 189.218 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19541-95-8
Molecular Formula C10H11N3O
Molecular Weight (g/mol) 189.218
MDL Number MFCD00462192
InChI Key UPAGEJODHNVJNM-UHFFFAOYSA-N
Synonym 3-4-methoxyphenyl-1h-pyrazol-5-amine, 5-4-methoxyphenyl-1h-pyrazol-3-amine, 3-amino-5-4-methoxyphenyl pyrazole, 5-amino-3-4-methoxyphenyl pyrazole, 3-amino-5-4-methoxyphenyl-1h-pyrazole, 5-4-methoxy-phenyl-2h-pyrazol-3-ylamine, 5-amino-3-4-methoxyphenyl-1h-pyrazole, 1h-pyrazol-3-amine, 5-4-methoxyphenyl, 5-4-methoxyphenyl-2h-pyrazol-3-amine, 5-4-methoxyphenyl-2h-pyrazol-3-ylamine
PubChem CID 2756469
IUPAC Name 5-(4-methoxyphenyl)-1H-pyrazol-3-amine
SMILES COC1=CC=C(C=C1)C2=CC(=NN2)N

Specifications

Melting Point 141°C to 143°C
Quantity 1 g
Formula Weight 189.22
Percent Purity 97%
Chemical Name or Material 5-Amino-3-(4-methoxyphenyl)-1H-pyrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only