missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-3-(2-thienyl)-1H-pyrazole, 98%

Catalog No. AAL0675203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0675203 1 g
AAL0675206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0675203 Supplier Thermo Scientific Chemicals Supplier No. L0675203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 96799-03-0 | C7H7N3S | 165.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 96799-03-0
Molecular Formula C7H7N3S
Molecular Weight (g/mol) 165.21
MDL Number MFCD00051815
InChI Key TXSOLYKLZBJHFF-UHFFFAOYSA-N
Synonym 5-amino-3-2-thienyl pyrazole, 3-thiophen-2-yl-1h-pyrazol-5-amine, 5-thien-2-yl-1h-pyrazol-3-amine, 3-2-thienyl-1h-pyrazol-5-amine, 5-2-thienyl-1h-pyrazol-3-amine, 5-thiophen-2-yl-2h-pyrazol-3-ylamine, 5-thiophen-2-yl-1h-pyrazol-3-amine, gnf-pf-1850, 5-amino-3-thien-2-yl pyrazole, 5-amino-3-2-thienyl-1h-pyrazole
PubChem CID 523184
IUPAC Name 5-thiophen-2-yl-1H-pyrazol-3-amine
SMILES NC1=NNC(=C1)C1=CC=CS1

Specifications

Melting Point 101°C to 103°C
Quantity 1 g
Beilstein 4399374
Formula Weight 165.21
Percent Purity 98%
Chemical Name or Material 5-Amino-3-(2-thienyl)-1H-pyrazole
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only